Learn More
Medchemexpress LLC Doxazosin-d8 hydrochloride | 1219803-95-8 | 99.5% | C23H18D8ClN5O5 | 1 MG
SDP
Catalog No. 5000432681
Encompass_Preferred
Doxazosin-d8 hydrochloride is the deuterium labeled Doxazosin hydrochloride. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32681 Supplier Medchemexpress LLC Supplier No. HYB0098BS1MG
Doxazosin-d8 hydrochloride is the deuterium labeled Doxazosin hydrochloride. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.