Learn More
Medchemexpress LLC Doxazosin-d8 hydrochloride | 1219803-95-8 | 99.5% | C23H18D8ClN5O5 | 5 MG
SDP
Catalog No. 5000432306
Encompass_Preferred
Doxazosin-d8 (hydrochloride) is the deuterium-labeled Doxazosin hydrochloride. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32306 Supplier Medchemexpress LLC Supplier No. HYB0098BS5MG
Doxazosin-d8 (hydrochloride) is the deuterium-labeled Doxazosin hydrochloride. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.