Learn More
Apexbio Technology LLC Doxazosin Mesylate, 50mg. Cas: 77883-43-3 MFCD: MFCD00216023. α1-adrenergic receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A288450
Encompass_Preferred
Doxazosin Mesylate is a selective, long-lasting antagonistof1-adrenergic receptor [1].Doxazosin Mesylate has been reported to potentiatecontraction response to 5-HT in a dose-dependent manner [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4764