Learn More
Cayman Chemical Doxazosin (mesylate), Purity: ≥98%, CAS: 77883-43-3, 100 mg
SDP
Catalog No. 502967049
Encompass_Preferred
An α1-AR antagonist (Kis = 3.16, 1, and 3.98 nM for α1A­-, α1B-, and α1D-ARs, respectively); inhibits norepinephrine-induced contractions in isolated rat aorta rings and human prostate strips (pA2s = 8.8 and 8.2, respectively); inhibits phenylephrine-induced increases in blood pressure and prostatic pressure in anesthetized dogs (pA2 = 7.5 for both)
Catalog No. 50-296-7049 Supplier Cayman Chemical Supplier No. 18633100 MG
An α1-AR antagonist (Kis = 3.16, 1, and 3.98 nM for α1A­-, α1B-, and α1D-ARs, respectively); inhibits norepinephrine-induced contractions in isolated rat aorta rings and human prostate strips (pA2s = 8.8 and 8.2, respectively); inhibits phenylephrine-induced increases in blood pressure and prostatic pressure in anesthetized dogs (pA2 = 7.5 for both)