Learn More
Selleck Chemical LLC Doxepin E7725-100MG
SDP
Catalog No. 5000042065
Encompass_Preferred
Doxepin is a potent inhibitor of serotonin reuptake and noradrenaline reuptake with a Ki of 68 nM and 29 5 nM respectively It acts as a receptor antagonist for histamine H1 acetylcholine muscarinic and 1 adrenoceptor with a Ki of 0 24 nM 83 nM and 24 nM respectively and significantly inhibits CYP450
Catalog No. 50-000-42065 Supplier Selleck Chemical LLC Supplier No. E7725100MG
Doxepin is a potent inhibitor of serotonin reuptake and noradrenaline reuptake with a Ki of 68 nM and 29 5 nM respectively It acts as a receptor antagonist for histamine H1 acetylcholine muscarinic and 1 adrenoceptor with a Ki of 0 24 nM 83 nM and 24 nM respectively and significantly inhibits CYP450