Learn More
Medchemexpress LLC Doxepin (Hydrochloride) | 1229-29-4 | 99.6% | 1 G
SDP
Catalog No. 5000408940
Encompass_Preferred
Doxepin hydrochloride is an orally active tricyclic antidepressant agent that functions as a potent and selective histamine receptor H1 antagonist. It also significantly inhibits CYP450 2C19 and 1A2 and inhibits the reuptake of serotonin and norepinephrine. Its therapeutic effects include treating atopic dermatitis and chronic urticarial, improving cognitive processes, and protecting the central nervous system.
  • Potent and selective histamine receptor H1 antagonist
  • Potent CYP450 inhibitor
  • Inhibits reuptake of serotonin and norepinephrine
  • Supports treatment for atopic dermatitis and chronic urticarial
  • Improves cognitive processes and protects the central nervous system
Catalog No. 50-004-08940 Supplier Medchemexpress LLC Supplier No. HYB07251G
Doxepin hydrochloride is an orally active tricyclic antidepressant agent that functions as a potent and selective histamine receptor H1 antagonist. It also significantly inhibits CYP450 2C19 and 1A2 and inhibits the reuptake of serotonin and norepinephrine. Its therapeutic effects include treating atopic dermatitis and chronic urticarial, improving cognitive processes, and protecting the central nervous system.
  • Potent and selective histamine receptor H1 antagonist
  • Potent CYP450 inhibitor
  • Inhibits reuptake of serotonin and norepinephrine
  • Supports treatment for atopic dermatitis and chronic urticarial
  • Improves cognitive processes and protects the central nervous system