Learn More
Medchemexpress LLC HY-B0725 500mg Medchemexpress, Doxepin (Hydrochloride) CAS:1229-29-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0725500MG
Encompass_Preferred
Medchemexpress, HY-B0725 500mg Doxepin (Hydrochloride) CAS:1229-29-4 Doxepin hydrochloride is an orally active tricyclic antidepressant agent. Doxepin hydrochloride is a potent and selective histamine receptor H1 antagonist. Doxepin hydrochloride is also a potent CYP450 inhibitor and significantly inhibits CYP450 2C19 and 1A2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2649