Learn More
Apexbio Technology LLC Doxepin (hydrochloride) 1229-29-4 5g
SDP
Catalog No. 502240365
Encompass_Preferred
Doxepin (hydrochloride) (CAS 1229-29-4) is a tricyclic antidepressant that functions as a potent antagonist of the histamine H1 receptor a G protein coupled receptor expressed in tissues such as smooth muscle vascular endothelium cardiac tissue and the central nervous system In vitro doxepin acts as a competitive inhibitor of serotonin uptake in human platelets with a reported Ki of approximately 0 2 M and can induce rapid serotonin efflux at higher concentrations In vivo studies in rodents and canines demonstrate efficient oral absorption and metabolic conversion in liver and urine with central nervous system penetration observed in rats Doxepin is widely used in research related to neurotransmitter modulation and histaminergic signaling
This item is not returnable. View return policy
Catalog No. 50-224-0365 Supplier Apexbio Technology LLC Supplier No. C41535000
Doxepin (hydrochloride) (CAS 1229-29-4) is a tricyclic antidepressant that functions as a potent antagonist of the histamine H1 receptor a G protein coupled receptor expressed in tissues such as smooth muscle vascular endothelium cardiac tissue and the central nervous system In vitro doxepin acts as a competitive inhibitor of serotonin uptake in human platelets with a reported Ki of approximately 0 2 M and can induce rapid serotonin efflux at higher concentrations In vivo studies in rodents and canines demonstrate efficient oral absorption and metabolic conversion in liver and urine with central nervous system penetration observed in rats Doxepin is widely used in research related to neurotransmitter modulation and histaminergic signaling