Learn More
Description
Doxepin Hydrochloride USP is a white or almost white crystalline hygroscopic powder
Specifications
Specifications
| Chemical Name or Material | Doxepin Hydrochloride USP |
| Packaging | Jar 500ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 1229-29-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 315.84 |
| Quantity | 100 g |
| Percent Purity | 98 to 102 % (Total doxepin hydrochloride) |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
