Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Doxepin Hydrochloride USP
SDP

Catalog No. 14186447 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-447 1 kg
14-185-424 100 g
14-186-433 500 g
14-185-432 5 g
14-185-376 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-186-447 Supplier Medisca Supplier No. 021809
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doxepin Hydrochloride USP is a white or almost white crystalline hygroscopic powder

Doxepin Hydrochloride USP is a white or almost white crystalline hygroscopic powder

Specifications

Chemical Name or Material Doxepin Hydrochloride USP
Packaging Jar 3800ml, White, HDPE, Round, 110-400, TRIVENI-DB, CT
CAS 1229-29-4
Type Drug Standard
Molecular Weight (g/mol) 315.84
Quantity 1 kg
Percent Purity 98 to 102 % (Total doxepin hydrochloride)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.