Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxepin Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602325 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-325 1 g Amber Glass Bottle
18-602-326 5 g Amber Glass Bottle
18-602-327 25 g Amber Glass Bottle
18-602-328 100 g Amber Glass Bottle
18-602-329 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-325 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D11951GM
Add to Cart
Add to Cart

D1195, 1229-29-4, C19H21NO·HCl

Spectrum™ Chemical Doxepin Hydrochloride, USP is a tricyclic antidepressant used to treat mental and mood problems such as depression and anxiety. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1229-29-4
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.84
InChI Key MHNSPTUQQIYJOT-SJDTYFKWSA-N
IUPAC Name hydrogen dimethyl({3-[(2E)-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene]propyl})amine chloride
SMILES [H+].[Cl-].CN(C)CC\C=C1/C2=CC=CC=C2COC2=CC=CC=C12

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.