Learn More
Cayman Chemical DoxepIn N-oxIdehydrate 10mg
SDP
Catalog No. 502893038
Encompass_Preferred
A metabolite of doxepin; a photodegradation product of doxepin
Catalog No. 50-289-3038 Supplier Cayman Chemical Supplier No. 900230210 MG
A metabolite of doxepin; a photodegradation product of doxepin