Learn More
TARGETMOL CHEMICALS INC DOXERCALCIFEROL 10MG
SDP
Catalog No. 502576600
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Doxercalciferol (TSA 840) is a synthetic vitamin D2 analog suppressing parathyroid synthesis and secretion used to treat secondary hyperparathyroidism and metabolic bone disease. purity: 99%
Catalog No. 50-257-6600 Supplier TARGETMOL CHEMICALS INC Supplier No. T611210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Doxercalciferol (TSA 840) is a synthetic vitamin D2 analog suppressing parathyroid synthesis and secretion used to treat secondary hyperparathyroidism and metabolic bone disease. purity: 99%