Learn More
Apexbio Technology LLC Doxercalciferol 54573-75-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061806
Encompass_Preferred
Doxercalciferol chemically known as 1 -Hydroxyvitamin D2 is a synthetic analog of vitamin D2 characterized by selective activation of the vitamin D receptor (VDR) Upon metabolic activation Doxercalciferol undergoes hepatic transformation to its biologically active form inducing gene transcription mediated via the VDR signaling pathway Central to its mechanism activated VDR regulates calcium and phosphate homeostasis influencing cellular differentiation proliferation and parathyroid hormone expression In biomedical research Doxercalciferol serves as a tool compound to investigate the molecular pathways regulated by vitamin D receptor activation particularly in renal insufficiency models bone metabolism studies and parathyroid physiology experiments
Catalog No. 50-000-61806 Supplier Apexbio Technology LLC Supplier No. B20915.1
Doxercalciferol chemically known as 1 -Hydroxyvitamin D2 is a synthetic analog of vitamin D2 characterized by selective activation of the vitamin D receptor (VDR) Upon metabolic activation Doxercalciferol undergoes hepatic transformation to its biologically active form inducing gene transcription mediated via the VDR signaling pathway Central to its mechanism activated VDR regulates calcium and phosphate homeostasis influencing cellular differentiation proliferation and parathyroid hormone expression In biomedical research Doxercalciferol serves as a tool compound to investigate the molecular pathways regulated by vitamin D receptor activation particularly in renal insufficiency models bone metabolism studies and parathyroid physiology experiments