Learn More
Medchemexpress LLC HY-32348 5mg Medchemexpress, Doxercalciferol CAS:54573-75-0 Purity:>98%
SDP
Catalog No. 502082229
Encompass_Preferred
Medchemexpress, HY-32348 5mg Doxercalciferol CAS:54573-75-0 Doxercalciferol is a Vitamin D2 analog, acts as an activator of Vitamin D receptor, and prevent renal disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2229 Supplier Medchemexpress LLC Supplier No. HY323485MG
Add to Cart
Add to Cart
Medchemexpress, HY-32348 5mg Doxercalciferol CAS:54573-75-0 Doxercalciferol is a Vitamin D2 analog, acts as an activator of Vitamin D receptor, and prevent renal disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.