Learn More
Selleck Chemical LLC Doxorubicin E2516-100MG
SDP
Catalog No. 5000040787
Encompass_Preferred
Adriamycin (Doxorubicin Hydroxydaunorubicin) a cytotoxic anthracycline antibiotic is an anti-cancer chemotherapy agent inhibits topoisomerase II with an IC50 of 2 67 M thus stopping DNA replication and induces apoptosis also inhibits human DNA topoisomerase I with an IC50 of 0 8 M
Catalog No. 50-000-40787 Supplier Selleck Chemical LLC Supplier No. E2516100MG
Adriamycin (Doxorubicin Hydroxydaunorubicin) a cytotoxic anthracycline antibiotic is an anti-cancer chemotherapy agent inhibits topoisomerase II with an IC50 of 2 67 M thus stopping DNA replication and induces apoptosis also inhibits human DNA topoisomerase I with an IC50 of 0 8 M