Learn More
Apexbio Technology LLC Doxorubicin 23214-92-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064318
Encompass_Preferred
Doxorubicin is an anthracycline antibiotic commonly used as an anti-cancer agent targeting DNA replication machinery Its mechanism of action involves intercalation into DNA double helices and subsequent inhibition of DNA topoisomerase II impeding DNA synthesis and causing genomic instability Additionally Doxorubicin mediates chromatin remodeling by facilitating histone displacement from active chromatin contributing further to DNA damage and transcriptional dysregulation It is extensively employed in studies involving hematologic malignancies solid tumors and sarcomas frequently serving as a chemotherapeutic reference compound In biochemical assays Doxorubicin has demonstrated inhibitory effects on Topoisomerase II with an IC50 typically ranging between 1-10 M depending on assay conditions and cell lines applied
Catalog No. 50-000-64318 Supplier Apexbio Technology LLC Supplier No. A39665.1
Doxorubicin is an anthracycline antibiotic commonly used as an anti-cancer agent targeting DNA replication machinery Its mechanism of action involves intercalation into DNA double helices and subsequent inhibition of DNA topoisomerase II impeding DNA synthesis and causing genomic instability Additionally Doxorubicin mediates chromatin remodeling by facilitating histone displacement from active chromatin contributing further to DNA damage and transcriptional dysregulation It is extensively employed in studies involving hematologic malignancies solid tumors and sarcomas frequently serving as a chemotherapeutic reference compound In biochemical assays Doxorubicin has demonstrated inhibitory effects on Topoisomerase II with an IC50 typically ranging between 1-10 M depending on assay conditions and cell lines applied