Learn More
Selleck Chemical LLC Doxorubicin-5mg
SDP
Catalog No. 502389704
Encompass_Preferred
Adriamycin (Doxorubicin, Hydroxydaunorubicin), a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent, inhibits topoisomerase II with an IC50 of 2.67 μM, thus stopping DNA replication, and induces apoptosis, also inhibits human DNA topoisomerase I with an IC50 of 0.8 μM.
Catalog No. 50-238-9704 Supplier Selleck Chemical LLC Supplier No. E25165MG
Adriamycin (Doxorubicin, Hydroxydaunorubicin), a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent, inhibits topoisomerase II with an IC50 of 2.67 μM, thus stopping DNA replication, and induces apoptosis, also inhibits human DNA topoisomerase I with an IC50 of 0.8 μM.