Learn More
Selleck Chemical LLC Doxorubicin (DOX) HCl S1208-1g
SDP
Catalog No. 503111417
Encompass_Preferred
Doxorubicin (DOX) HCl is an antibiotic agent that inhibits human DNA topoisomerase I and topoisomerase II with IC50s of 0 8 M and 2 67 M respectively Doxorubicin reduces basal phosphorylation of AMPK Doxorubicin is used in the concomitant treatment of HIV-infected patients but is found to be at high risk of HBV reactivation This product may precipitate when dissolved in PBS solution It is recommended to prepare the stock solution in pure water and dilute with either pure water or saline to obtain the working solution Doxorubicin (Adriamycin) HCl can be used to induce animal models of kidney disease
Catalog No. 50-311-1417 Supplier Selleck Chemical LLC Supplier No. S12081G
Doxorubicin (DOX) HCl is an antibiotic agent that inhibits human DNA topoisomerase I and topoisomerase II with IC50s of 0 8 M and 2 67 M respectively Doxorubicin reduces basal phosphorylation of AMPK Doxorubicin is used in the concomitant treatment of HIV-infected patients but is found to be at high risk of HBV reactivation This product may precipitate when dissolved in PBS solution It is recommended to prepare the stock solution in pure water and dilute with either pure water or saline to obtain the working solution Doxorubicin (Adriamycin) HCl can be used to induce animal models of kidney disease