Learn More
Apexbio Technology LLC Doxorubicin (Adriamycin) HCl, 100mg. Cas: 25316-40-9 MFCD: MFCD00077757. Antitumour antibiotic,inhibits TOPO II.
SDP
Catalog No. 501904711
Encompass_Preferred
Doxorubicin is a semi-synthesized anticancer agent derived from bacterial culture. [1] It is an anthracycline antibiotic. It is been widely used in blood cancers, solid tumors and sarcomas. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4711 Supplier Apexbio Technology LLC Supplier No. A1832100
Doxorubicin is a semi-synthesized anticancer agent derived from bacterial culture. [1] It is an anthracycline antibiotic. It is been widely used in blood cancers, solid tumors and sarcomas. Other sizes are available. Please inqury us for quote.