Learn More
Selleck Chemical LLC Doxorubicin (Adriamycin) HCl 10mg 25316-40-9 NSC 123127, DOX
SDP
Catalog No. NC0444049
Encompass_Preferred
Doxorubicin (Adriamycin, NSC 123127, DOX) HCl is an antibiotic agent that inhibits DNA topoisomerase II and induces DNA damage, mitophagy and apoptosis in tumor cells. Doxorubicin reduces basal phosphorylation of AMPK. Doxorubicin is used in the concomitant treatment of HIV-infected patients but is found to be at high risk of HBV reactivation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0444049 Supplier Selleck Chemical LLC Supplier No. S120810MG
Doxorubicin (Adriamycin, NSC 123127, DOX) HCl is an antibiotic agent that inhibits DNA topoisomerase II and induces DNA damage, mitophagy and apoptosis in tumor cells. Doxorubicin reduces basal phosphorylation of AMPK. Doxorubicin is used in the concomitant treatment of HIV-infected patients but is found to be at high risk of HBV reactivation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.