Learn More
Enzo Life Sciences Doxorubicin . HCl, (5mg), CAS Number: 25316-40-9
SDP
Catalog No. NC0652400
Encompass_Preferred
Topoisomerase inhibitor. A broad spectrum antitumor agent. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Binds covalently to DNA. Inhbibits reverse transcriptase, RNA polymerase and the catalytic activity of Dnmt1. Immunosuppressive. Antineoplastic. Induces apoptosis. Down regulates c-myc and c-jun in doxorubicin-sensitive cells. Alternative Name: DXR . HCl, 14-Hydroxydaunomycin . HCl, Adriamycin . HCl. Formula: C27H29NO11 . HCl. MW: 580.0. CAS Number: 25316-40-9. MI: 14: 3439. Purity: ≥95% (HPLC). Appearance: Red solid. Solubility: Soluble in water. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at room temperature. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: Ambient
Catalog No. NC0652400 Supplier Enzo Life Sciences Supplier No. BMLGR3190005
Topoisomerase inhibitor. A broad spectrum antitumor agent. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Binds covalently to DNA. Inhbibits reverse transcriptase, RNA polymerase and the catalytic activity of Dnmt1. Immunosuppressive. Antineoplastic. Induces apoptosis. Down regulates c-myc and c-jun in doxorubicin-sensitive cells. Alternative Name: DXR . HCl, 14-Hydroxydaunomycin . HCl, Adriamycin . HCl. Formula: C27H29NO11 . HCl. MW: 580.0. CAS Number: 25316-40-9. MI: 14: 3439. Purity: ≥95% (HPLC). Appearance: Red solid. Solubility: Soluble in water. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at room temperature. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: Ambient