Learn More
Apexbio Technology LLC Doxorubicin (Adriamycin) HCl 25316-40-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063696
Encompass_Preferred
Doxorubicin hydrochloride an anthracycline antibiotic derivative exhibits cytotoxic activity and is widely utilized as an anticancer chemotherapeutic agent Its mechanism involves intercalating into DNA double-strands inhibiting DNA topoisomerase II activity and consequently interrupting replication processes causing DNA damage and altered chromatin structure by histone displacement In biomedical research settings doxorubicin hydrochloride is employed in vitro and in animal models to study therapeutic strategies against hematologic malignancies solid tumors and sarcomas Reported IC50 values vary with cell type and assay conditions typically ranging between approximately 0 1 M to 2 M in various tumor-derived cell models
Catalog No. 50-000-63696 Supplier Apexbio Technology LLC Supplier No. A18325.1
Doxorubicin hydrochloride an anthracycline antibiotic derivative exhibits cytotoxic activity and is widely utilized as an anticancer chemotherapeutic agent Its mechanism involves intercalating into DNA double-strands inhibiting DNA topoisomerase II activity and consequently interrupting replication processes causing DNA damage and altered chromatin structure by histone displacement In biomedical research settings doxorubicin hydrochloride is employed in vitro and in animal models to study therapeutic strategies against hematologic malignancies solid tumors and sarcomas Reported IC50 values vary with cell type and assay conditions typically ranging between approximately 0 1 M to 2 M in various tumor-derived cell models