Learn More
Medchemexpress LLC HY-15142 500mg , Doxorubicin (hydrochloride) CAS:25316-40-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15142500MG
Encompass_Preferred
HY-15142 500mg Doxorubicin (hydrochloride) CAS: 25316-40-9 Doxorubicin (Hydroxydaunorubicin) hydrochloride, a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent. Doxorubicin hydrochloride is a potent human DNA topoisomerase I and topoisomerase II inhibitor with IC50s of 0.8 μM and 2.67 μM, respectively. Doxorubicin hydrochloride reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Doxorubicin hydrochloride induces apoptosis and autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9104