Learn More
Ubpbio Doxorubicin HCL, 5MG
Catalog No. 501965139
Encompass_Preferred
Doxorubicin is an anthracycline antitumor antibiotic that Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Doxorubicin binds covalently to DNA and inhbibits reverse transcriptase, RNA polymerase and the catalytic activity of Dnmt1.
Catalog No. 50-196-5139 Supplier Ubpbio Supplier No. F8110
Doxorubicin is an anthracycline antitumor antibiotic that Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Doxorubicin binds covalently to DNA and inhbibits reverse transcriptase, RNA polymerase and the catalytic activity of Dnmt1.