Learn More
Bioworld Doxorubicin HCl, 10 mg
Supplier:  Bioworld 404100041
Encompass_Preferred
Doxorubicin, HCl Antineoplastic antibiotic interferes with nucleic acid synthesis. Also acts on cell membranes.Light/moisture sensitiveA Topo II inhibitor, DNA intercalator and powerful cytotoxic agentPossible risk of irreversible effects.For in vitro lab use only. Not for human use or consumption.Usage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: SolidBoiling Point : 810.3 °C at 760 mmHgMelting Point: 203.9-205 °CRTECS Number: QI9295900
Catalog No. 50-255-867