Learn More
Research Products International Corp Doxorubicin HCl Powder 5 MG
SDP
Catalog No. 501977873
Encompass_Preferred
Doxorubicin Hydrochloride is a potent chemotherapeutic agent. It intercalates with DNA, inhibiting nucleic acid synthesis, and also generates free radicals that cause DNA strand breaks. These actions disrupt the replication and transcription processes, leading to cell death. It also generates free radicals that can cause DNA damage and induce apoptosis (programmed cell death) in cancer cells.
Doxorubicin is widely used in cancer research as a tool to study cellular responses to DNA damage, apoptosis, and mechanisms of drug resistance.
WARNING! This product can expose you to Doxorubicin HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7873 Supplier Research Products International Corp Supplier No. D430100.005
Doxorubicin Hydrochloride is a potent chemotherapeutic agent. It intercalates with DNA, inhibiting nucleic acid synthesis, and also generates free radicals that cause DNA strand breaks. These actions disrupt the replication and transcription processes, leading to cell death. It also generates free radicals that can cause DNA damage and induce apoptosis (programmed cell death) in cancer cells.
Doxorubicin is widely used in cancer research as a tool to study cellular responses to DNA damage, apoptosis, and mechanisms of drug resistance.
WARNING! This product can expose you to Doxorubicin HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.