Learn More
Medchemexpress LLC HY-15142 100mg Medchemexpress, Doxorubicin (hydrochloride) CAS:25316-40-9 Purity:>98%
SDP
Catalog No. 502029081
Encompass_Preferred
Medchemexpress, HY-15142 100mg Doxorubicin (hydrochloride) CAS:25316-40-9 Doxorubicin hydrochloride (Hydroxydaunorubicin hydrochloride), a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent. Doxorubicin hydrochloride inhibits topoisomerase II with an IC50 of 2.67 μM, thus stopping DNA replication. Doxorubicin hydrochloride reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Doxorubicin hydrochloride induces apoptosis and autophagy[2]. Doxorubicin hydrochloride inhibits human DNA topoisomerase I with an IC50 of 0.8 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9081 Supplier Medchemexpress LLC Supplier No. HY15142100MG
Medchemexpress, HY-15142 100mg Doxorubicin (hydrochloride) CAS:25316-40-9 Doxorubicin hydrochloride (Hydroxydaunorubicin hydrochloride), a cytotoxic anthracycline antibiotic, is an anti-cancer chemotherapy agent. Doxorubicin hydrochloride inhibits topoisomerase II with an IC50 of 2.67 μM, thus stopping DNA replication. Doxorubicin hydrochloride reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Doxorubicin hydrochloride induces apoptosis and autophagy[2]. Doxorubicin hydrochloride inhibits human DNA topoisomerase I with an IC50 of 0.8 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.