Learn More
Medchemexpress LLC Doxorubicin hydrochloride | 25316-40-9 | C₂₇H₃₀ClNO₁₁ | 5 MG
SDP
Catalog No. 5000372193
Encompass_Preferred
Doxorubicin hydrochloride is a cytotoxic anthracycline antibiotic and an anti-cancer chemotherapy agent. It is a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. It also induces apoptosis and autophagy.
  • Potent human DNA topoisomerase I and topoisomerase II inhibitor
  • Reduces basal phosphorylation of AMPK
  • Induces apoptosis and autophagy
  • Used in infection and cancer research
Catalog No. 50-003-72193 Supplier Medchemexpress LLC Supplier No. HY15142R5MG
Doxorubicin hydrochloride is a cytotoxic anthracycline antibiotic and an anti-cancer chemotherapy agent. It is a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. It also induces apoptosis and autophagy.
  • Potent human DNA topoisomerase I and topoisomerase II inhibitor
  • Reduces basal phosphorylation of AMPK
  • Induces apoptosis and autophagy
  • Used in infection and cancer research