Learn More
Medchemexpress LLC Doxorubicin (hydrochloride) | 25316-40-9 | C27H30ClNO11 | 100 MG
SDP
Catalog No. 5000372457
Encompass_Preferred
Doxorubicin hydrochloride (Standard) is an analytical standard and cytotoxic anthracycline antibiotic, primarily used as an anti-cancer chemotherapy agent. It functions as a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase, while also inducing apoptosis and autophagy.
  • Intended for research and analytical applications
  • Functions as a cytotoxic anthracycline antibiotic
  • Acts as an anti-cancer chemotherapy agent
  • Potent human DNA topoisomerase I and II inhibitor
  • Induces apoptosis and autophagy
  • Used in qualitative and quantitative research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-72457 Supplier Medchemexpress LLC Supplier No. HY15142R100MG
Doxorubicin hydrochloride (Standard) is an analytical standard and cytotoxic anthracycline antibiotic, primarily used as an anti-cancer chemotherapy agent. It functions as a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase, while also inducing apoptosis and autophagy.
  • Intended for research and analytical applications
  • Functions as a cytotoxic anthracycline antibiotic
  • Acts as an anti-cancer chemotherapy agent
  • Potent human DNA topoisomerase I and II inhibitor
  • Induces apoptosis and autophagy
  • Used in qualitative and quantitative research experiments
  • Suitable for HPLC, GC, and MS applications