Learn More
Medchemexpress LLC Doxorubicin (hydrochloride) | 25316-40-9 | C27H30ClNO11 | 25 MG
SDP
Catalog No. 5000374522
Encompass_Preferred
Doxorubicin hydrochloride (Hydroxydaunorubicin hydrochloride; ADR) is a cytotoxic anthracycline antibiotic and an anti-cancer chemotherapy agent. It functions as a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Additionally, it induces apoptosis and autophagy.
  • Cytotoxic anthracycline antibiotic
  • Anti-cancer chemotherapy agent
  • Potent human DNA topoisomerase I and II inhibitor
  • Reduces basal phosphorylation of AMPK and acetyl-CoA carboxylase
  • Induces apoptosis and autophagy
  • Suitable for neutral or weakly acidic solvents
  • Melting/freezing point of 216°C
  • For research use only
Catalog No. 50-003-74522 Supplier Medchemexpress LLC Supplier No. 25MGHY15142R
Doxorubicin hydrochloride (Hydroxydaunorubicin hydrochloride; ADR) is a cytotoxic anthracycline antibiotic and an anti-cancer chemotherapy agent. It functions as a potent human DNA topoisomerase I and topoisomerase II inhibitor, reducing basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase. Additionally, it induces apoptosis and autophagy.
  • Cytotoxic anthracycline antibiotic
  • Anti-cancer chemotherapy agent
  • Potent human DNA topoisomerase I and II inhibitor
  • Reduces basal phosphorylation of AMPK and acetyl-CoA carboxylase
  • Induces apoptosis and autophagy
  • Suitable for neutral or weakly acidic solvents
  • Melting/freezing point of 216°C
  • For research use only