Learn More
Medchemexpress LLC Doxorubicin hydrochloride | 25316-40-9 | C27H30ClNO11 | 50 MG
SDP
Catalog No. 5000373456
Encompass_Preferred
Doxorubicin hydrochloride is an analytical standard and a cytotoxic anthracycline antibiotic. This anti-cancer chemotherapy agent is a potent human DNA topoisomerase I and topoisomerase II inhibitor. It induces apoptosis and autophagy and is used for research and analytical applications.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments
  • Potent human DNA topoisomerase I and topoisomerase II inhibitor
  • Reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase
  • Induces apoptosis and autophagy
Catalog No. 50-003-73456 Supplier Medchemexpress LLC Supplier No. HY15142R50MG
Doxorubicin hydrochloride is an analytical standard and a cytotoxic anthracycline antibiotic. This anti-cancer chemotherapy agent is a potent human DNA topoisomerase I and topoisomerase II inhibitor. It induces apoptosis and autophagy and is used for research and analytical applications.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments
  • Potent human DNA topoisomerase I and topoisomerase II inhibitor
  • Reduces basal phosphorylation of AMPK and its downstream target acetyl-CoA carboxylase
  • Induces apoptosis and autophagy