Learn More
Medchemexpress LLC Doxorubicin hydrochloride | 25316-40-9 | MFCD00077757 | 99.3% | 579.98 g/mol | C27H30ClNO11 | 10 MG
SDP
Catalog No. 5000286460
Encompass_Preferred
Doxorubicin hydrochloride is an anthracycline antibiotic with cytotoxic activity supplied as a high-purity analytical standard for laboratory research. It functions as a DNA intercalator and inhibits topoisomerases I and II, making it useful as a biochemical reagent and reference in cell-based assays and analytical method development.
  • High purity (99.33%) suitable for analytical applications.
  • Confirmed identity: CAS 25316-40-9, molecular weight 579.98 g/mol.
  • Available in multiple package sizes, including 10 mg.
  • Acts as a DNA intercalator and topoisomerase I/II inhibitor.
  • Suitable for biochemical assays, cell-based studies, and analytical standards.
Catalog No. 50-002-86460 Supplier Medchemexpress LLC Supplier No. HY15142R10MG
Doxorubicin hydrochloride is an anthracycline antibiotic with cytotoxic activity supplied as a high-purity analytical standard for laboratory research. It functions as a DNA intercalator and inhibits topoisomerases I and II, making it useful as a biochemical reagent and reference in cell-based assays and analytical method development.
  • High purity (99.33%) suitable for analytical applications.
  • Confirmed identity: CAS 25316-40-9, molecular weight 579.98 g/mol.
  • Available in multiple package sizes, including 10 mg.
  • Acts as a DNA intercalator and topoisomerase I/II inhibitor.
  • Suitable for biochemical assays, cell-based studies, and analytical standards.