Learn More
Medchemexpress LLC HY-15142 1g , Doxorubicin (hydrochloride) Adriamycin;Hydroxydaunorubicin hydrochloride CAS:25316-40-9 Purity:98%
SDP
Catalog No. 501492273
Encompass_Preferred
Medchemexpress, HY-15142 1g Doxorubicin (hydrochloride) Adriamycin;Hydroxydaunorubicin hydrochloride CAS:25316-40-9 Formula:C27H30ClNO11 Topoisomerase II Purity:98% Doxorubicin hydrochloride is a Topoisomerase II (Top2) catalytic inhibitor, also is a broad spectrum antibiotic used in the treatment of cancers, with IC 50 of 374 nM for Hela cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2273 Supplier Medchemexpress LLC Supplier No. HY151421G
Medchemexpress, HY-15142 1g Doxorubicin (hydrochloride) Adriamycin;Hydroxydaunorubicin hydrochloride CAS:25316-40-9 Formula:C27H30ClNO11 Topoisomerase II Purity:98% Doxorubicin hydrochloride is a Topoisomerase II (Top2) catalytic inhibitor, also is a broad spectrum antibiotic used in the treatment of cancers, with IC 50 of 374 nM for Hela cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.