Learn More
Alkali Scientific Doxorubicin Hydrochloride, Powder, 10 Mg
SDP
Catalog No. 5000661001
Encompass_Preferred
Doxorubicin Hydrochloride (powder form, 10 mg) is a chemotherapy drug used in the treatment of various cancers such as leukemia, breast cancer, and sarcomas. As a potent anthracycline antibiotic, it interferes with DNA replication and transcription by inserting itself between DNA base pairs, effectively halting cell division. This powder form allows for precise reconstitution, offering flexibility in clinical and research applications. Doxorubicin is crucial in cancer treatment protocols, often used in combination with other drugs to enhance therapeutic outcomes. Its cytotoxic action on cancer cells makes it one of the most widely utilized chemotherapeutic agents in oncological care.
Catalog No. 50-006-61001 Supplier Alkali Scientific Supplier No. OXO002CP
Doxorubicin Hydrochloride (powder form, 10 mg) is a chemotherapy drug used in the treatment of various cancers such as leukemia, breast cancer, and sarcomas. As a potent anthracycline antibiotic, it interferes with DNA replication and transcription by inserting itself between DNA base pairs, effectively halting cell division. This powder form allows for precise reconstitution, offering flexibility in clinical and research applications. Doxorubicin is crucial in cancer treatment protocols, often used in combination with other drugs to enhance therapeutic outcomes. Its cytotoxic action on cancer cells makes it one of the most widely utilized chemotherapeutic agents in oncological care.