Learn More
Alkali Scientific Doxorubicin Hydrochloride, Powder, 5 Milligrams
SDP
Catalog No. 5000660081
Encompass_Preferred
Doxorubicin Hydrochloride, 5mg is an anticancer drug used in laboratory research for testing and studying chemotherapy treatments. Doxorubicin is an anthracycline antibiotic that intercalates into DNA, inhibiting the enzyme topoisomerase II and disrupting DNA replication, which leads to cell death. This compound is widely used in cancer research to explore its effectiveness in various cancer cell lines. The 5mg vial is suitable for preparing solutions for in vitro studies of cell growth inhibition, apoptosis, and drug resistance. It is an essential reagent in preclinical and clinical research investigating cancer therapies and chemotherapy resistance mechanisms.
Catalog No. 50-006-60081 Supplier Alkali Scientific Supplier No. OXO004CP
Doxorubicin Hydrochloride, 5mg is an anticancer drug used in laboratory research for testing and studying chemotherapy treatments. Doxorubicin is an anthracycline antibiotic that intercalates into DNA, inhibiting the enzyme topoisomerase II and disrupting DNA replication, which leads to cell death. This compound is widely used in cancer research to explore its effectiveness in various cancer cell lines. The 5mg vial is suitable for preparing solutions for in vitro studies of cell growth inhibition, apoptosis, and drug resistance. It is an essential reagent in preclinical and clinical research investigating cancer therapies and chemotherapy resistance mechanisms.