Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Doxorubicin-SMCC 10mM 1mL | 400647-59-8 | 1 ML
SDP

Catalog No. 5000229732
Encompass_Preferred

Doxorubicin-SMCC is a payload-linker conjugate that links the DNA topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker for antibody-drug conjugate research. It is supplied as a 10 mM solution in DMSO.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Non-cleavable SMCC linker for stable maleimide-thiol conjugation.
  • Doxorubicin payload (topoisomerase II inhibitor) for potent cytotoxicity.
  • High purity (98.8%) for reproducible conjugation and bioassays.
  • Molecular weight 762.76 g·mol-1; formula C39H42N2O14.
  • Storage: in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-29732 Supplier Medchemexpress LLC Supplier No. HY11606310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doxorubicin-SMCC is a payload-linker conjugate that links the DNA topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker for antibody-drug conjugate research. It is supplied as a 10 mM solution in DMSO.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Non-cleavable SMCC linker for stable maleimide-thiol conjugation.
  • Doxorubicin payload (topoisomerase II inhibitor) for potent cytotoxicity.
  • High purity (98.8%) for reproducible conjugation and bioassays.
  • Molecular weight 762.76 g·mol-1; formula C39H42N2O14.
  • Storage: in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.