Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Doxorubicin-SMCC 25mg | 400647-59-8 | 762.76 | C39H42N2O14 | 25 MG
SDP

Catalog No. 5000227237
Encompass_Preferred

Doxorubicin-SMCC is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples the anthracycline topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker, producing a stable payload for bioconjugation and preclinical studies.

  • Non-cleavable SMCC linker for stable conjugation.
  • Acts as a DNA topoisomerase II inhibitor payload.
  • Purity 98.8% and molecular weight 762.76.
  • Orange to red solid physical form.
  • Soluble in DMSO (125 mg/mL); ultrasonic recommended.
  • CAS number 400647-59-8 for cross-referencing.

Catalog No. 50-002-27237 Supplier Medchemexpress LLC Supplier No. HY11606325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doxorubicin-SMCC is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples the anthracycline topoisomerase II inhibitor doxorubicin to a non-cleavable SMCC linker, producing a stable payload for bioconjugation and preclinical studies.

  • Non-cleavable SMCC linker for stable conjugation.
  • Acts as a DNA topoisomerase II inhibitor payload.
  • Purity 98.8% and molecular weight 762.76.
  • Orange to red solid physical form.
  • Soluble in DMSO (125 mg/mL); ultrasonic recommended.
  • CAS number 400647-59-8 for cross-referencing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.