Learn More
Cayman Chemical Doxorubicin (hydrochloride), Purity: ≥98%, CAS: 25316-40-9, 50 mg
SDP
Catalog No. 502969546
Encompass_Preferred
An anthracycline antitumor antibiotic agent that inhibits DNA topoisomerase II by inducing double-stranded DNA breaks; inhibits nucleic acid synthesis and induces apoptosis by intercalating within DNA
Catalog No. 50-296-9546 Supplier Cayman Chemical Supplier No. 1500750 MG
An anthracycline antitumor antibiotic agent that inhibits DNA topoisomerase II by inducing double-stranded DNA breaks; inhibits nucleic acid synthesis and induces apoptosis by intercalating within DNA