Learn More
Cayman Chemical Doxorubicinol (hydrochloride), Purity: ≥90% (mixture of isomers), 5 mg
SDP
Catalog No. 502986781
Encompass_Preferred
A doxorubicin metabolite; formed by NADPH-dependent reduction of the side chain carbonyl group of doxorubicin in human, rabbit, and canine cardiac tissue; inhibits sodium-potassium-dependent ATPase activity (IC50 = 5.40 μg/ml) and ATP-dependent calcium uptake (IC50= 4.5 μg/ml) in canine cardiac muscle; increases resting stress in contracting rabbit cardiac muscle
Catalog No. 50-298-6781 Supplier Cayman Chemical Supplier No. 223865 MG
A doxorubicin metabolite; formed by NADPH-dependent reduction of the side chain carbonyl group of doxorubicin in human, rabbit, and canine cardiac tissue; inhibits sodium-potassium-dependent ATPase activity (IC50 = 5.40 μg/ml) and ATP-dependent calcium uptake (IC50= 4.5 μg/ml) in canine cardiac muscle; increases resting stress in contracting rabbit cardiac muscle