Learn More
Formumax Scientific Inc DOXOVES-PEGYL LD HCL 2ML, 4MG
Catalog No. NC1488582
Encompass_Preferred
Doxoves® - PEGylated Liposomal Doxorubicin HCl (2.0mL, 4.0mg/mL)Doxorubicin HCl is loaded into PEGylated liposomes composed of HSPC/CHOL/mPEG2000-DSPE (56.2:38.5:5.3 mol/mol) with ammonium gradient by the active loading approach. It is comparable to the commercial product DOXIL®. The liposomes are prepared to very high quality with consistent lot-lot characteristics. Narrow particle size distribution and high drug encapsulation efficiency. Note the high drug concentration (4mg/mL compared to 2 mg/mL for DOXIL®) allows the researchers to perform experiments at higher concentration/doses.Control liposomes for this product: F30204B-C-2 (please inquire)Product For Research only and NOT for Human Use
Catalog No. NC1488582 Supplier Formumax Scientific Inc Supplier No. F30204BD2
Doxoves® - PEGylated Liposomal Doxorubicin HCl (2.0mL, 4.0mg/mL)Doxorubicin HCl is loaded into PEGylated liposomes composed of HSPC/CHOL/mPEG2000-DSPE (56.2:38.5:5.3 mol/mol) with ammonium gradient by the active loading approach. It is comparable to the commercial product DOXIL®. The liposomes are prepared to very high quality with consistent lot-lot characteristics. Narrow particle size distribution and high drug encapsulation efficiency. Note the high drug concentration (4mg/mL compared to 2 mg/mL for DOXIL®) allows the researchers to perform experiments at higher concentration/doses.Control liposomes for this product: F30204B-C-2 (please inquire)Product For Research only and NOT for Human Use