Learn More
Medchemexpress LLC HY-N0565 100mg , Doxycycline CAS:564-25-0 Purity:>98%
SDP
Catalog No. 502261547
Encompass_Preferred
HY-N0565 100mg Doxycycline CAS: 564-25-0 Doxycycline, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline shows antibacterial activity and anti-cancer cell proliferation activity[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1547 Supplier Medchemexpress LLC Supplier No. HYN0565100MG
HY-N0565 100mg Doxycycline CAS: 564-25-0 Doxycycline, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline shows antibacterial activity and anti-cancer cell proliferation activity[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.