Learn More
TARGETMOL CHEMICALS INC Doxycycline 200MG
SDP
Catalog No. 502877383
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Doxycycline (Doxiciclina) belongs to the tetracycline class of antibiotics and is a broad-spectrum metalloproteinase (MMP) inhibitor with oral activity. Doxycycline has antibacterial activity and antitumor activity. Purity 99.88%
Catalog No. 50-287-7383 Supplier TARGETMOL CHEMICALS INC Supplier No. T1687200MG
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Doxycycline (Doxiciclina) belongs to the tetracycline class of antibiotics and is a broad-spectrum metalloproteinase (MMP) inhibitor with oral activity. Doxycycline has antibacterial activity and antitumor activity. Purity 99.88%