Learn More
Selleck Chemical LLC Doxycycline 25mg 564-25-0 Vibramycin, Doxytetracycline, Doxiciclina, Doxycyclinum
SDP
Catalog No. 501948062
Encompass_Preferred
Doxycycline (Vibramycin, Doxytetracycline, Doxiciclina, Doxycyclinum) is an antibiotic that is used in the treatment of a number of types of infections caused by bacteria and protozoa. Doxycycline is also a nonspecific matrix metalloproteinase (MMP) inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8062 Supplier Selleck Chemical LLC Supplier No. S515925MG
Doxycycline (Vibramycin, Doxytetracycline, Doxiciclina, Doxycyclinum) is an antibiotic that is used in the treatment of a number of types of infections caused by bacteria and protozoa. Doxycycline is also a nonspecific matrix metalloproteinase (MMP) inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.