Learn More
Medchemexpress LLC Doxycycline | 564-25-0 | MFCD00800994 | 444.4 g/mol | 100 MG
SDP
Catalog No. 5000388559
Encompass_Preferred
Doxycycline is a semi-synthetic tetracycline antibiotic that functions by inhibiting bacterial protein synthesis. It is widely used to treat various bacterial infections and also exhibits geroprotector, antimalarial, immunomodulatory, and anti-inflammatory properties.
  • Inhibits bacterial protein synthesis
  • Treats bacterial infections
  • Acts as a geroprotector
  • Possesses antimalarial properties
  • Functions as an immunomodulator
  • Exhibits anti-inflammatory effects
Catalog No. 50-003-88559 Supplier Medchemexpress LLC Supplier No. HYN0565R100MG
Doxycycline is a semi-synthetic tetracycline antibiotic that functions by inhibiting bacterial protein synthesis. It is widely used to treat various bacterial infections and also exhibits geroprotector, antimalarial, immunomodulatory, and anti-inflammatory properties.
  • Inhibits bacterial protein synthesis
  • Treats bacterial infections
  • Acts as a geroprotector
  • Possesses antimalarial properties
  • Functions as an immunomodulator
  • Exhibits anti-inflammatory effects