Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Doxycycline | 564-25-0 | MFCD00800994 | 97.2% | 444.43 | C22H24N2O8 | 10 MG
SDP

Catalog No. 5000296856
Encompass_Preferred

Doxycycline analytical standard is an analytical reference material of the tetracycline antibiotic doxycycline intended for research and analytical applications. It is supplied as a solid reference for assay calibration, method development, and quality control in laboratories.

  • Analytical standard for assay calibration and method validation.
  • Intended for research and analytical applications.
  • Reported purity 97.2%.
  • CAS number 564-25-0 for substance identification.
  • Chemical formula C22H24N2O8; molecular weight 444.43 g/mol.
  • Available in small reference quantities such as 10 mg for analytical use.

Catalog No. 50-002-96856 Supplier Medchemexpress LLC Supplier No. HYN0565R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Doxycycline analytical standard is an analytical reference material of the tetracycline antibiotic doxycycline intended for research and analytical applications. It is supplied as a solid reference for assay calibration, method development, and quality control in laboratories.

  • Analytical standard for assay calibration and method validation.
  • Intended for research and analytical applications.
  • Reported purity 97.2%.
  • CAS number 564-25-0 for substance identification.
  • Chemical formula C22H24N2O8; molecular weight 444.43 g/mol.
  • Available in small reference quantities such as 10 mg for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.