Learn More
Medchemexpress LLC Doxycycline-d3 (hyclate) (major) | 564-25-0 | 1 MG
SDP
Catalog No. 5000388556
Encompass_Preferred
Doxycycline-d3 hyclate (major) is the deuterium labeled Doxycycline hyclate (major) (HY-N0565). It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88556 Supplier Medchemexpress LLC Supplier No. HYN0565S11MG
Doxycycline-d3 hyclate (major) is the deuterium labeled Doxycycline hyclate (major) (HY-N0565). It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.