Learn More
Med Vet International Zoetis Doxirobe Gel, 8.5%, 3 Syringe/Box
Catalog No. 502828148
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Doxirobe Gel (doxycycline 8.5%) is indicated for the treatment and control of periodontal disease in dogs. Doxycycline is a semi-synthetic tetracycline derivative that has a wide range of antimicrobial activity against microorganisms, particularly Gram-negative anaerobic bacteria, involved in periodontal disease. Upon contact with the aqueous environment (gingival crevicular fluid) the polymer will coagulate, resulting in the formation of a solid, pliable delivery system within the treated periodontal pocket(s). Doxycycline is released into the gingival crevicular fluid for a local effect on the microorganisms. Do not use in dogs younger than 1 year, the use of tetracyclines during tooth development has been associated with permanent discoloration of the teeth. Do not use in pregnant bitches. Clients should be advised to suspend brushing treated teeth for approximately 2 weeks following treatment.
Catalog No. 50-282-8148 Supplier Med Vet International Supplier No. RXDOXIROBE
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Doxirobe Gel (doxycycline 8.5%) is indicated for the treatment and control of periodontal disease in dogs. Doxycycline is a semi-synthetic tetracycline derivative that has a wide range of antimicrobial activity against microorganisms, particularly Gram-negative anaerobic bacteria, involved in periodontal disease. Upon contact with the aqueous environment (gingival crevicular fluid) the polymer will coagulate, resulting in the formation of a solid, pliable delivery system within the treated periodontal pocket(s). Doxycycline is released into the gingival crevicular fluid for a local effect on the microorganisms. Do not use in dogs younger than 1 year, the use of tetracyclines during tooth development has been associated with permanent discoloration of the teeth. Do not use in pregnant bitches. Clients should be advised to suspend brushing treated teeth for approximately 2 weeks following treatment.