Learn More
Bioworld Doxycycline HCl, 10 g
Catalog No. 50255868
Encompass_Preferred
Doxycycline HClActive against Gram-positive and Gram-negative bacteriaInterferes with bacterial cell wall synthesisSemi-synthetic tetracyclineSynonym: Synonym: 2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aR,5S,5aR,6R,12aS)- SpecificationsLight sensitiveUsage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: Solid
Catalog No. 50-255-868 Supplier Bioworld Supplier No. 404100051
Doxycycline HClActive against Gram-positive and Gram-negative bacteriaInterferes with bacterial cell wall synthesisSemi-synthetic tetracyclineSynonym: Synonym: 2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aR,5S,5aR,6R,12aS)- SpecificationsLight sensitiveUsage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: Solid