Learn More
Med Vet International Doxycycline Hyclate Tablets (Oracea), 100mg, 50ct
Catalog No. 502828150
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Doxycycline hyclate tablets is a generic version of Oracea. It belongs to a class of antibiotics known as tetracyclines. It should be stored at room temperature between 59-86 degrees F and away from light and moisture.
Catalog No. 50-282-8150 Supplier Med Vet International Supplier No. RX27808023401
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Doxycycline hyclate tablets is a generic version of Oracea. It belongs to a class of antibiotics known as tetracyclines. It should be stored at room temperature between 59-86 degrees F and away from light and moisture.